accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3CCH3OOHNBOHHO

CAS RN 489446-42-6

IUPAC Name: [4-({[(tert-butoxy)carbonyl]amino}methyl)phenyl]boronic acid
Synonyms: 4-{[(tert-butoxycarbonyl)amino]methyl}phenylboronic acid
Molecular Weight (g/mol): 251.09
Molecular Formula: C12H18BNO4
InChi Key: MUBGEKQUCSEECZ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results