accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 487-90-1

IUPAC Name: 3-[5-(aminomethyl)-4-(carboxymethyl)-1H-pyrrol-3-yl]propanoic acid
Synonyms: porphobilinogen pbg - porphobilinogen porphobilinogen (substance)
Molecular Weight (g/mol): 226.23
Molecular Formula: C10H14N2O4
InChi Key: QSHWIQZFGQKFMA-UHFFFAOYSA-N
SMILES: NCC1=C(CC(O)=O)C(CCC(O)=O)=CN1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)