accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 486-73-7

HOON

CAS RN 486-73-7

IUPAC Name: isoquinoline-1-carboxylic acid
Synonyms: isoquinoline-1-carboxylic acid
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: XAAKCCMYRKZRAK-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C=CC=CC2=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results