accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4856-95-5

ONHBH3

CAS RN 4856-95-5

IUPAC Name: boron(3+) morpholine trihydride
Synonyms: boron(3+) morpholine trihydride
Molecular Weight (g/mol): 100.96
Molecular Formula: C4H12BNO
InChi Key: WILVPUFGQGZGOC-UHFFFAOYSA-N
SMILES: [H-].[H-].[H-].[B+3].C1COCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results