accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4837-01-8

BrS

CAS RN 4837-01-8

IUPAC Name: [(2-bromoethyl)sulfanyl]benzene
Synonyms: [(2-bromoethyl)sulfanyl]benzene
Molecular Weight (g/mol): 217.12
Molecular Formula: C8H9BrS
InChi Key: UEFBOQYLXLEJSM-UHFFFAOYSA-N
SMILES: BrCCSC1=CC=CC=C1
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)