accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BrOOCH2

CAS RN 4823-47-6

IUPAC Name: 2-bromoethyl prop-2-enoate
Synonyms: 2-bromoethyl prop-2-enoate
Molecular Weight (g/mol): 179.01
Molecular Formula: C5H7BrO2
InChi Key: CDZAAIHWZYWBSS-UHFFFAOYSA-N
SMILES: BrCCOC(=O)C=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results