accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COHCH2

CAS RN 4798-44-1

IUPAC Name: hex-1-en-3-ol
Synonyms: 1-hexen-3-ol
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: BVOSSZSHBZQJOI-UHFFFAOYNA-N
SMILES: CCCC(O)C=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results