accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4771-49-7

CH3HNO

CAS RN 4771-49-7

IUPAC Name: 6-methyl-1H-indole-3-carbaldehyde
Synonyms: 6-methyl-1H-indole-3-carbaldehyde
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: LZERQSJGPXFAKB-UHFFFAOYSA-N
SMILES: CC1=CC=C2C(NC=C2C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results