accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClSN

CAS RN 4771-31-7

IUPAC Name: 4-(chloromethyl)-2-phenyl-1,3-thiazole
Synonyms: 4-(chloromethyl)-2-phenyl-1,3-thiazole
Molecular Weight (g/mol): 209.69
Molecular Formula: C10H8ClNS
InChi Key: SVEGSFSFMLCNFF-UHFFFAOYSA-N
SMILES: ClCC1=CSC(=N1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results