accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 476004-82-7

BOOH3CH3CCH3H3CNSiH3CCH3CH3CH3H3CH3C

CAS RN 476004-82-7

IUPAC Name: 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-[tris(propan-2-yl)silyl]-1H-indole
Synonyms: 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(triisopropylsilyl)indole
Molecular Weight (g/mol): 399.46
Molecular Formula: C23H38BNO2Si
InChi Key: MXFFSQNKCJHCKO-UHFFFAOYSA-N
SMILES: CC(C)[Si](C(C)C)(C(C)C)N1C=C(B2OC(C)(C)C(C)(C)O2)C2=CC=CC=C12
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results