CAS RN 4755-81-1
CAS RN 4755-81-1
IUPAC Name:
methyl 2-chloro-3-oxobutanoate
Synonyms:
methyl 2-chloro-3-oxobutanoate
Molecular Weight (g/mol):
150.56
Molecular Formula:
C5H7ClO3
InChi Key:
GYQRIAVRKLRQKP-UHFFFAOYNA-N
SMILES:
COC(=O)C(Cl)C(C)=O
1
–
1
of
1
results
1
Methyl 2-chloroacetoacetate, 95%
CAS: 4755-81-1 Molecular Formula: C5H7ClO3 Molecular Weight (g/mol): 150.56 MDL Number: MFCD00008757 InChI Key: GYQRIAVRKLRQKP-UHFFFAOYSA-N Synonym: methyl 2-chloroacetoacetate,butanoic acid, 2-chloro-3-oxo-, methyl ester,2-chloroacetoacetic acid methyl ester,2-chloro-3-oxo-butyric acid methyl ester,methyl2-chloroacetoacetate,acmc-209k9v,methyl-2-chloroacetoacetate,methyl-alpha-chloroacetoacetate,2-chloroacetoacetate methyl ester,gyqriavrklrqkp-uhfffaoysa PubChem CID: 107332 IUPAC Name: methyl 2-chloro-3-oxobutanoate SMILES: CC(=O)C(C(=O)OC)Cl