accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4721-24-8

H3CPOOO

CAS RN 4721-24-8

IUPAC Name: hexylphosphonate
Synonyms: hexylphosphonate
Molecular Weight (g/mol): 164.14
Molecular Formula: C6H13O3P
InChi Key: GJWAEWLHSDGBGG-UHFFFAOYSA-L
SMILES: CCCCCCP([O-])([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results