accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4707-24-8

HOOFFBrFF

CAS RN 4707-24-8

IUPAC Name: 4-bromo-2,3,5,6-tetrafluorobenzoic acid
Synonyms: 4-bromo-2,3,5,6-tetrafluorobenzoic acid
Molecular Weight (g/mol): 272.98
Molecular Formula: C7HBrF4O2
InChi Key: MTZLICDXLGQWJV-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C(F)=C(Br)C(F)=C1F
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results