accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3CH3H3COONNBOH3CH3CH3CCH3O

CAS RN 470478-90-1

IUPAC Name: tert-butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine-1-carboxylate
Synonyms: tert-butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine-1-carboxylate
Molecular Weight (g/mol): 388.32
Molecular Formula: C21H33BN2O4
InChi Key: ZMAVVXGWEHZLDW-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results