accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4695-13-0

H2NO

CAS RN 4695-13-0

IUPAC Name: 2,2-diphenylacetamide
Synonyms: acetamide, 2,2-diphenyl-
Molecular Weight (g/mol): 211.26
Molecular Formula: C14H13NO
InChi Key: ZXQVXEAZKZFEEP-UHFFFAOYSA-N
SMILES: NC(=O)C(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results