accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CPOHOHO

CAS RN 4671-75-4

IUPAC Name: tetradecylphosphonic acid
Synonyms: phosphonic acid, tetradecyl-
Molecular Weight (g/mol): 278.37
Molecular Formula: C14H31O3P
InChi Key: BVQJQTMSTANITJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCP(O)(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results