accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3ONHHO

CAS RN 4664-16-8

IUPAC Name: 6-hydroxy-4-methyl-1,2-dihydropyridin-2-one
Synonyms: 6-hydroxy-4-methyl-1H-pyridin-2-one
Molecular Weight (g/mol): 125.13
Molecular Formula: C6H7NO2
InChi Key: JJHVYGVVMBYCMQ-UHFFFAOYSA-N
SMILES: CC1=CC(=O)NC(O)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results