accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ONHON

CAS RN 4664-01-1

IUPAC Name: 1H,2H,3H-pyrrolo[3,4-c]pyridine-1,3-dione
Synonyms: 2H-pyrrolo[3,4-c]pyridine-1,3-dione
Molecular Weight (g/mol): 148.12
Molecular Formula: C7H4N2O2
InChi Key: SJSABZBUTDSWMJ-UHFFFAOYSA-N
SMILES: O=C1NC(=O)C2=CN=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results