accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 465514-03-8

IUPAC Name: 3-(4-methoxyphenyl)-5-methyl-1,2-oxazole-4-carbonyl chloride
Synonyms: 3-(4-methoxyphenyl)-5-methyl-1,2-oxazole-4-carbonyl chloride
Molecular Weight (g/mol): 251.67
Molecular Formula: C12H10ClNO3
InChi Key: FABGSPUDPOKWCR-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=NOC(C)=C1C(Cl)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results