accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4630-82-4

CH3OO

CAS RN 4630-82-4

IUPAC Name: methyl cyclohexanecarboxylate
Synonyms: cyclohexanecarboxylic acid, methyl ester
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: ZQWPRMPSCMSAJU-UHFFFAOYSA-N
SMILES: COC(=O)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results