accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4629-50-9

CH3NH3COOONCH3H3C

CAS RN 4629-50-9

IUPAC Name: 3-(dimethylamino)benzoyl 3-(dimethylamino)benzoate
Synonyms: 3-(dimethylamino)benzoyl 3-(dimethylamino)benzoate
Molecular Weight (g/mol): 312.37
Molecular Formula: C18H20N2O3
InChi Key: ROXBMXDDDMXDFQ-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=CC(=C1)C(=O)OC(=O)C1=CC(=CC=C1)N(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results