CAS RN 462-34-0
CAS RN 462-34-0
IUPAC Name:
oxolane; trifluoroborane
Synonyms:
THF; boron trifluoride
Molecular Weight (g/mol):
139.91
Molecular Formula:
C4H8BF3O
InChi Key:
CHNLPLHJUPMEOI-UHFFFAOYSA-N
SMILES:
FB(F)F.C1CCOC1
1
–
2
of
2
results
1
Promotions Available
Boron trifluoride-tetrahydrofuran complex, BF{3} 45.5%
Boron trifluoride-tetrahydrofuran complex, BF{3} 45.5%
CAS: 462-34-0 Molecular Formula: C4H8BF3O Molecular Weight (g/mol): 139.91 MDL Number: MFCD00040372 InChI Key: CHNLPLHJUPMEOI-UHFFFAOYSA-N Synonym: boron trifluoride tetrahydrofuran complex,trifluoro tetrahydro-1h-furan-1-ium-1-yl borate,pubchem7062,trifluoro oxolan-1-ium-1-yl boranuide,borontrifluoride tetrahydrofuran complex,boron trifluoride tetrahydrofuran complex, contains sulfur dioxide as peroxide formation suppressor PubChem CID: 60146129 SMILES: FB(F)F.C1CCOC1