CAS RN 4612-28-6
CAS RN 4612-28-6
IUPAC Name:
[3-(dihydroxyboranyl)phenyl]boronic acid
Synonyms:
3-(dihydroxyboranyl)phenylboronic acid
Molecular Weight (g/mol):
165.75
Molecular Formula:
C6H8B2O4
InChi Key:
UXPAASVRXBULRG-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC(=CC=C1)B(O)O
1
–
1
of
1
results
1
1,3-Benzenediboronic acid, 97%, Thermo Scientific™
CAS: 4612-28-6 Molecular Formula: C6H8B2O4 Molecular Weight (g/mol): 165.746 MDL Number: MFCD03092915 InChI Key: UXPAASVRXBULRG-UHFFFAOYSA-N Synonym: 1,3-benzenediboronic acid,1,3-phenylenediboronic acid,1,3-benzendiboronic acid,1,3-benzenediboronicacid,m-phenylenediboronic acid,3-dihydroxyboranyl phenyl boronic acid,3-dihydroxyboranyl phenylboronic acid,m-benzenediboronic acid,pubchem17415,1,3-benzendibororic acid PubChem CID: 230477 IUPAC Name: (3-boronophenyl)boronic acid SMILES: B(C1=CC(=CC=C1)B(O)O)(O)O