accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NH2NH3CN

CAS RN 461-98-3

IUPAC Name: 2,6-dimethylpyrimidin-4-amine
Synonyms: 2,6-dimethyl-4-pyrimidinamine
Molecular Weight (g/mol): 123.16
Molecular Formula: C6H9N3
InChi Key: BJJDXAFKCKSLTE-UHFFFAOYSA-N
SMILES: CC1=CC(N)=NC(C)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results