accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H2NSNN

CAS RN 4604-72-2

IUPAC Name: pyrazine-2-carbothioamide
Synonyms: pyrazinecarboxamide, thio-
Molecular Weight (g/mol): 139.18
Molecular Formula: C5H5N3S
InChi Key: LIURPUMROGYCLW-UHFFFAOYSA-N
SMILES: NC(=S)C1=CN=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results