accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4597-87-9

H3CNHN

CAS RN 4597-87-9

IUPAC Name: N-methylpyridin-2-amine
Synonyms: 2-methylaminopyridine
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: SVEUVITYHIHZQE-UHFFFAOYSA-N
SMILES: CNC1=CC=CC=N1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)