accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CONNFBFFF

CAS RN 459-64-3

IUPAC Name: boron(3+) 4-methoxybenzene-1-diazonium tetrafluoride
Synonyms: boron(3+) 4-methoxybenzenediazonium tetrafluoride
Molecular Weight (g/mol): 221.95
Molecular Formula: C7H7BF4N2O
InChi Key: CACFTKIREZJSIG-UHFFFAOYSA-J
SMILES: [B+3].[F-].[F-].[F-].[F-].COC1=CC=C(C=C1)[N+]#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results