CAS RN 458-50-4
CAS RN 458-50-4
IUPAC Name:
1-bromo-2-fluoro-4-methoxybenzene
Synonyms:
1-bromo-2-fluoro-4-methoxybenzene
Molecular Weight (g/mol):
205.03
Molecular Formula:
C7H6BrFO
InChi Key:
XANVIFOBBVAKCY-UHFFFAOYSA-N
SMILES:
COC1=CC(F)=C(Br)C=C1
1
–
1
of
1
results
1
4-Bromo-3-fluoroanisole, 97%, Thermo Scientific™
CAS: 458-50-4 Molecular Formula: C7H6BrFO Molecular Weight (g/mol): 205.026 MDL Number: MFCD01310983 InChI Key: XANVIFOBBVAKCY-UHFFFAOYSA-N Synonym: 4-bromo-3-fluoroanisole,3-fluoro-4-bromo anisole,3-fluoro-4-bromoanisol,3-fluoro-4-bromo anisol,1-bromo-2-fluoro-4-methoxy-benzene,benzene, 1-bromo-2-fluoro-4-methoxy,pharmabridge p-1018,rarechem al mz 0889,zlchem 365,pubchem1970 PubChem CID: 3718444 IUPAC Name: 1-bromo-2-fluoro-4-methoxybenzene SMILES: COC1=CC(=C(C=C1)Br)F