accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 457628-14-7

H3COFOOCH3

CAS RN 457628-14-7

IUPAC Name: 6-fluoro-2,3-dimethoxybenzaldehyde
Synonyms: 6-fluoro-2,3-dimethoxybenzaldehyde
Molecular Weight (g/mol): 184.17
Molecular Formula: C9H9FO3
InChi Key: GDOQJEBNBOJBQX-UHFFFAOYSA-N
SMILES: COC1=C(OC)C(C=O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results