accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CBrNH2

CAS RN 45762-41-2

IUPAC Name: 4-bromo-2-ethylaniline
Synonyms: 4-bromo-2-ethylaniline
Molecular Weight (g/mol): 200.08
Molecular Formula: C8H10BrN
InChi Key: LGOZNQPHTIGMQJ-UHFFFAOYSA-N
SMILES: CCC1=C(N)C=CC(Br)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results