accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H2NSOO

CAS RN 4563-33-1

IUPAC Name: phenylmethanesulfonamide
Synonyms: benzenemethanesulfonamide
Molecular Weight (g/mol): 171.21
Molecular Formula: C7H9NO2S
InChi Key: ABOYDMHGKWRPFD-UHFFFAOYSA-N
SMILES: NS(=O)(=O)CC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results