accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFFS

CAS RN 456-56-4

IUPAC Name: [(trifluoromethyl)sulfanyl]benzene
Synonyms: trifluoromethylthiobenzene
Molecular Weight (g/mol): 178.17
Molecular Formula: C7H5F3S
InChi Key: YQQKTCBMKQQOSM-UHFFFAOYSA-N
SMILES: FC(F)(F)SC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results