accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4556-23-4

SNH

CAS RN 4556-23-4

IUPAC Name: 1,4-dihydropyridine-4-thione
Synonyms: 4(1H)-pyridinethione
Molecular Weight (g/mol): 111.16
Molecular Formula: C5H5NS
InChi Key: FHTDDANQIMVWKZ-UHFFFAOYSA-N
SMILES: S=C1C=CNC=C1
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results