accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4553-21-3

IUPAC Name: 4-(2-azaniumylethyl)-1-benzylpiperazine-1,4-diium
Synonyms: 4-(2-ammonioethyl)-1-benzylpiperazine-1,4-diium
Molecular Weight (g/mol): 222.35
Molecular Formula: C13H24N3
InChi Key: CXEJMFLWEVKOGS-UHFFFAOYSA-Q
SMILES: [NH3+]CC[NH+]1CC[NH+](CC2=CC=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results