accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 455280-00-9

IUPAC Name: 5-bromo-1,2-benzoxazol-3-amine
Synonyms: 5-bromo-1,2-benzoxazol-3-amine
Molecular Weight (g/mol): 213.03
Molecular Formula: C7H5BrN2O
InChi Key: BGMYWBQEKIMHGU-UHFFFAOYSA-N
SMILES: NC1=NOC2=CC=C(Br)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results