CAS RN 4551-69-3
CAS RN 4551-69-3
IUPAC Name:
4-benzoyl-3-methyl-1-phenyl-4,5-dihydro-1H-pyrazol-5-one
Synonyms:
4-benzoyl-5-methyl-2-phenyl-4H-pyrazol-3-one
Molecular Weight (g/mol):
278.31
Molecular Formula:
C17H14N2O2
InChi Key:
JDOLPAMOKSAEMD-UHFFFAOYNA-N
SMILES:
CC1=NN(C(=O)C1C(=O)C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
4-Benzoyl-3-methyl-1-phenyl-5-pyrazolinone, 98+%, Thermo Scientific Chemicals
CAS: 4551-69-3 Molecular Formula: C17H14N2O2 MDL Number: MFCD00003136 Synonym: 4-Benzoyl-2,4-dihydro-5-methyl-2-phenyl-3H-pyrazol-3-one