accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 455-86-7

HOOFF

CAS RN 455-86-7

IUPAC Name: 3,4-difluorobenzoic acid
Synonyms: 3,4-fifluorobenzoic acid
Molecular Weight (g/mol): 158.1
Molecular Formula: C7H4F2O2
InChi Key: FPENCTDAQQQKNY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(F)C(F)=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results