accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFFN

CAS RN 455-18-5

IUPAC Name: 4-(trifluoromethyl)benzonitrile
Synonyms: 4-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 171.12
Molecular Formula: C8H4F3N
InChi Key: DRNJIKRLQJRKMM-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)C#N
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)