accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4548-45-2

ONOClN

CAS RN 4548-45-2

IUPAC Name: 2-chloro-5-nitropyridine
Synonyms: pyridine, 2-chloro-5-nitro-
Molecular Weight (g/mol): 158.54
Molecular Formula: C5H3ClN2O2
InChi Key: BAZVFQBTJPBRTJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Cl)N=C1
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)