accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4541-32-6

CH3H3CO

CAS RN 4541-32-6

IUPAC Name: 2,2-dimethylcyclopentan-1-one
Synonyms: 2,2-dimethylcyclopentan-1-one
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: FTGZMZBYOHMEPS-UHFFFAOYSA-N
SMILES: CC1(C)CCCC1=O

 No Refinements