accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2BrFFF

CAS RN 454-79-5

IUPAC Name: 2-bromo-5-(trifluoromethyl)aniline
Synonyms: 2-bromo-5-(trifluoromethyl)aniline
Molecular Weight (g/mol): 240.02
Molecular Formula: C7H5BrF3N
InChi Key: PZDVFXUBTKPFSG-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1Br)C(F)(F)F
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)