accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4530-18-1

H3CH3CCH3OONHOHO

CAS RN 4530-18-1

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-phenylpropanoic acid
Molecular Weight (g/mol): 265.31
Molecular Formula: C14H19NO4
InChi Key: ZYJPUMXJBDHSIF-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)