accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HOS

CAS RN 4521-31-7

IUPAC Name: [2-(hydroxymethyl)phenyl]sulfanide
Synonyms: [2-(hydroxymethyl)phenyl]sulfanide
Molecular Weight (g/mol): 139.19
Molecular Formula: C7H7OS
InChi Key: FYWFCRHZXORPFH-UHFFFAOYSA-M
SMILES: OCC1=CC=CC=C1[S-]
Boiling Point 
  • (3)
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)