accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NH2F

CAS RN 452-69-7

IUPAC Name: 4-fluoro-3-methylaniline
Synonyms: 4-fluoro-3-methylaniline
Molecular Weight (g/mol): 125.15
Molecular Formula: C7H8FN
InChi Key: NYMDPDNETOLVBS-UHFFFAOYSA-N
SMILES: CC1=CC(N)=CC=C1F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results