CAS RN 4509-90-4
CAS RN 4509-90-4
IUPAC Name:
N-[2-(pyridin-4-yl)-1,3-benzoxazol-5-yl]acetamide
Synonyms:
N-[2-(pyridin-4-yl)-1,3-benzoxazol-5-yl]acetamide
Molecular Weight (g/mol):
253.26
Molecular Formula:
C14H11N3O2
InChi Key:
QANQQXSQVQYQFI-UHFFFAOYSA-N
SMILES:
CC(=O)NC1=CC=C2OC(=NC2=C1)C1=CC=NC=C1
1
–
1
of
1
results
1
5-Bromovaleryl chloride, 98%
CAS: 4509-90-4 Molecular Formula: C14H11N3O2 Molecular Weight (g/mol): 253.26 MDL Number: MFCD00013660 InChI Key: QANQQXSQVQYQFI-UHFFFAOYSA-N Synonym: 5-bromovaleryl chloride,5-bromovaleroyl chloride,5-bromo-pentanoyl chloride,5-bromovaleric acid chloride,5-bromovalerylchloride,bromovalerylchloride,bromovaleryl chloride,5-bromovaleric chloride,5-bromopentanoylchloride,5-bromo pentanoyl chloride PubChem CID: 521173 IUPAC Name: 5-bromopentanoyl chloride SMILES: CC(=O)NC1=CC=C2OC(=NC2=C1)C1=CC=NC=C1