accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NNH3CCNNN

CAS RN 448245-52-1

IUPAC Name: 1-butyl-3-methyl-1H-imidazol-3-ium; [(cyanoimino)methylidene]azanide
Synonyms: 1-butyl-3-methylimidazolium; [(cyanoimino)methylidene]azanide
Molecular Weight (g/mol): 205.27
Molecular Formula: C10H15N5
InChi Key: ICIVTHOGIQHZRY-UHFFFAOYSA-N
SMILES: [N-]=C=NC#N.CCCCN1C=C[N+](C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results