CAS RN 4463-44-9
CAS RN 4463-44-9
IUPAC Name:
sodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-6-hydroxybenzene-1,3-disulfonic acid hydride
Synonyms:
sodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-6-hydroxybenzene-1,3-disulfonic acid hydride
Molecular Weight (g/mol):
556.62
Molecular Formula:
C25H29N2NaO7S2
InChi Key:
HLEQELPTFNOLFE-ZCTWALFCSA-N
SMILES:
[H-].[Na+].CCNC1=CC=C(C=C1C)C(=C1/C=CC(=NCC)C(C)=C1)\C1=CC(O)=C(C=C1S(O)(=O)=O)S(O)(=O)=O
1
–
1
of
1
results
1
Xylene Cyanol, pure
CAS: 4463-44-9 Molecular Formula: C25H28N2NaO7S2 Molecular Weight (g/mol): 555.616 MDL Number: MFCD00019481 InChI Key: JKPDSWLBCLGXEQ-UHFFFAOYSA-N Synonym: C.I. 43535,Cyanol FF PubChem CID: 131854036 IUPAC Name: 4-[[4-(ethylamino)-3-methylphenyl]-(4-ethylimino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-6-hydroxybenzene-1,3-disulfonic acid;sodium SMILES: CCNC1=C(C=C(C=C1)C(=C2C=CC(=NCC)C(=C2)C)C3=CC(=C(C=C3S(=O)(=O)O)S(=O)(=O)O)O)C.[Na]