accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4463-33-6

H3COH3COCH3

CAS RN 4463-33-6

IUPAC Name: 1,2-dimethoxy-3-methylbenzene
Synonyms: 1,2-dimethoxy-3-methyl-benzene
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: WMXFNCKPYCAIQW-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C)=C1OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results