CAS RN 446299-84-9
IUPAC Name:
N-(5-bromo-3-methylpyridin-2-yl)-2,2-dimethylpropanamide
Synonyms:
N-(5-bromo-3-methylpyridin-2-yl)-2,2-dimethylpropanamide
Molecular Weight (g/mol):
271.16
Molecular Formula:
C11H15BrN2O
InChi Key:
DJAJPKVIGCOGCY-UHFFFAOYSA-N
SMILES:
CC1=CC(Br)=CN=C1NC(=O)C(C)(C)C
1
–
1
of
1
results
1
5-Bromo-3-methyl-2-(2,2,2-trimethylacetamido)pyridine, 95%, Thermo Scientific™
CAS: 446299-84-9 Molecular Formula: C11H15BrN2O Molecular Weight (g/mol): 271.158 MDL Number: MFCD09037447 InChI Key: DJAJPKVIGCOGCY-UHFFFAOYSA-N Synonym: n-5-bromo-3-methylpyridin-2-yl pivalamide,5-bromo-2-pivaloylamino-3-picoline,n-5-bromo-3-methylpyridin-2-yl-2,2-dimethylpropanamide,5-bromo-3-methyl-2-2,2,2-trimethylacetamido pyridine,n-5-bromo-3-methyl-2-pyridyl pivalamide,5-bromo-3-methyl-2-pivaloylamino pyridine,n-5-bromo-3-methyl-2-pyridinyl-2,2-dimethylpropanamide PubChem CID: 42553022 IUPAC Name: N-(5-bromo-3-methylpyridin-2-yl)-2,2-dimethylpropanamide SMILES: CC1=CC(=CN=C1NC(=O)C(C)(C)C)Br