accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OF

CAS RN 446-22-0

IUPAC Name: 1-(2-fluorophenyl)propan-1-one
Synonyms: 1-(2-fluorophenyl)propan-1-one
Molecular Weight (g/mol): 152.17
Molecular Formula: C9H9FO
InChi Key: NSNSIFGTEGKZFK-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=CC=C1F
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)